The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198463.2 | NP_940865.1 | 13 - 14 | 1829 | 59 |
XM_005264929.2 | XP_005264986.1 | 14 - 15 | 2548 | 59 | |
XM_006713013.3 | XP_006713076.1 | 14 - 15 | 2587 | 59 | |
XM_011533451.2 | XP_011531753.1 | 13 - 14 | 2422 | 59 | |
XM_011533456.1 | XP_011531758.1 | 14 - 15 | 2296 | 59 | |
XM_011533457.1 | XP_011531759.1 | 12 - 13 | 2076 | 59 | |
XM_011533458.2 | XP_011531760.1 | 11 - 12 | 1959 | 59 | |
XM_011533459.2 | XP_011531761.1 | 13 - 14 | 2209 | 59 | |
XM_011533460.2 | XP_011531762.1 | 9 - 10 | 1726 | 59 | |
XM_011533461.1 | XP_011531763.1 | 11 - 12 | 1919 | 59 | |
XM_011533463.2 | XP_011531765.1 | 8 - 9 | 1550 | 59 | |
XM_017005848.1 | XP_016861337.1 | 10 - 11 | 1832 | 59 | |
XM_017005849.1 | XP_016861338.1 | 13 - 14 | 2430 | 59 | |
XM_017005850.1 | XP_016861339.1 | 12 - 13 | 1730 | 59 | |
GenBank mRNA | AK124111.1 | - | 13 - 14 | 1831 | 59 |
AK124920.1 | - | 6 - 7 | 1299 | 59 | |
AK128200.1 | - | 2 - 3 | 397 | 59 | |
BC132815.1 | - | 11 - 12 | 1643 | 59 | |
BC151142.1 | - | 11 - 12 | 1643 | 59 |