Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
Chromosome Location
Chr.3: 58716415 - 59050078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005264929.2 XP_005264986.1 12 - 13 1975 90
XM_006713013.3 XP_006713076.1 12 - 13 2014 90
XM_011533451.2 XP_011531753.1 11 - 12 1849 90
XM_011533453.2 XP_011531755.1 12 - 13 2014 90
XM_011533455.2 XP_011531757.1 12 - 13 2014 90
XM_011533456.1 XP_011531758.1 12 - 13 1723 90
XM_011533457.1 XP_011531759.1 10 - 11 1503 90
XM_011533458.2 XP_011531760.1 9 - 10 1386 90
XM_011533460.2 XP_011531762.1 7 - 8 1153 90
XM_011533461.1 XP_011531763.1 9 - 10 1346 90
XM_011533463.2 XP_011531765.1 6 - 7 977 90
XM_017005848.1 XP_016861337.1 8 - 9 1259 90
XM_017005849.1 XP_016861338.1 11 - 12 1857 90
GenBank mRNA AK124920.1 - 4 - 5 726 90
BC050317.1 - 12 - 13 2023 90

View Full Product Details