The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199022.2 | NP_001185951.2 | 1 - 2 | 233 | 149 |
NM_001323572.1 | NP_001310501.1 | 1 - 2 | 233 | 149 | |
NM_001323577.1 | NP_001310506.1 | 1 - 2 | 233 | 149 | |
GenBank mRNA | AK300426.1 | - | 1 - 2 | 146 | 149 |
BY798911.2 | - | 1 - 2 | 233 | 149 | |
CR749255.1 | - | 1 - 2 | 133 | 149 |