The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199022.2 | NP_001185951.2 | 7 - 8 | 2608 | 101 |
NM_001323569.1 | NP_001310498.1 | 8 - 9 | 2751 | 101 | |
NM_001323570.1 | NP_001310499.1 | 9 - 10 | 3196 | 101 | |
NM_001323571.1 | NP_001310500.1 | 9 - 10 | 2991 | 101 | |
NM_001323572.1 | NP_001310501.1 | 7 - 8 | 2608 | 101 | |
NM_001323576.1 | NP_001310505.1 | 8 - 9 | 2751 | 101 | |
NM_001323577.1 | NP_001310506.1 | 7 - 8 | 2608 | 101 | |
NM_014711.5 | NP_055526.4 | 8 - 9 | 2751 | 101 | |
XM_011545991.2 | XP_011544293.1 | 9 - 10 | 3174 | 101 | |
XM_011545992.2 | XP_011544294.1 | 9 - 10 | 2892 | 101 | |
XM_017023908.1 | XP_016879397.1 | 8 - 9 | 3668 | 101 | |
GenBank mRNA | AB007879.1 | - | 8 - 9 | 2652 | 101 |
AK300426.1 | - | 7 - 8 | 2521 | 101 | |
BC036654.2 | - | 8 - 9 | 2722 | 101 | |
CR749255.1 | - | 7 - 8 | 2508 | 101 | |
DA370828.1 | - | 2 - 3 | 174 | 101 |