The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321837.1 | NP_001308766.1 | 4 - 5 | 791 | 64 |
NM_001321838.1 | NP_001308767.1 | 4 - 5 | 791 | 64 | |
NM_004056.5 | NP_004047.3 | 4 - 5 | 791 | 64 | |
NR_135821.1 | 4 - 5 | 791 | 64 | ||
XM_017013818.1 | XP_016869307.1 | 5 - 6 | 856 | 64 | |
GenBank mRNA | AK090655.1 | - | 4 - 5 | 763 | 64 |
AK289470.1 | - | 4 - 5 | 763 | 64 | |
AK293999.1 | - | 3 - 4 | 382 | 64 | |
AK314538.1 | - | 4 - 5 | 764 | 64 | |
AY075022.1 | - | 4 - 5 | 768 | 64 | |
BC069744.1 | - | 4 - 5 | 760 | 64 | |
BC069794.1 | - | 4 - 5 | 795 | 64 | |
BC108929.2 | - | 4 - 5 | 716 | 64 | |
BM462961.1 | - | 4 - 5 | 764 | 64 | |
L04656.1 | - | 4 - 5 | 513 | 64 |