The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290474.1 | NP_001277403.1 | 1 - 2 | 745 | 66 |
NM_014807.4 | NP_055622.3 | 1 - 2 | 745 | 66 | |
XM_006718948.3 | XP_006719011.1 | 1 - 2 | 1034 | 66 | |
XM_017018625.1 | XP_016874114.1 | 1 - 2 | 1034 | 66 | |
XM_017018626.1 | XP_016874115.1 | 1 - 2 | 1034 | 66 | |
XM_017018627.1 | XP_016874116.1 | 1 - 2 | 1034 | 66 | |
GenBank mRNA | AB006623.2 | - | 1 - 2 | 210 | 66 |
AB094094.1 | - | 1 - 2 | 713 | 66 | |
AK025223.1 | - | 1 - 2 | 713 | 66 | |
BC022219.1 | - | 1 - 2 | 611 | 66 |