The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290474.1 | NP_001277403.1 | 5 - 6 | 1189 | 64 |
NM_014807.4 | NP_055622.3 | 5 - 6 | 1189 | 64 | |
XM_006718948.3 | XP_006719011.1 | 4 - 5 | 1367 | 64 | |
XM_011543076.2 | XP_011541378.1 | 5 - 6 | 1685 | 64 | |
XM_017018625.1 | XP_016874114.1 | 4 - 5 | 1367 | 64 | |
XM_017018626.1 | XP_016874115.1 | 5 - 6 | 1478 | 64 | |
XM_017018627.1 | XP_016874116.1 | 5 - 6 | 1478 | 64 | |
XM_017018628.1 | XP_016874117.1 | 5 - 6 | 614 | 64 | |
GenBank mRNA | AB006623.2 | - | 5 - 6 | 654 | 64 |
AB094094.1 | - | 5 - 6 | 1157 | 64 | |
AK025223.1 | - | 5 - 6 | 1157 | 64 | |
BC022219.1 | - | 5 - 6 | 1055 | 64 | |
BX537572.1 | - | 2 - 3 | 490 | 64 |