The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014366.4 | NP_055181.3 | 12 - 13 | 1495 | 85 |
NM_206825.1 | NP_996561.1 | 12 - 13 | 1592 | 85 | |
NM_206826.1 | NP_996562.1 | 12 - 13 | 1445 | 85 | |
GenBank mRNA | AF191018.1 | - | 12 - 13 | 1492 | 85 |
AK027514.1 | - | 12 - 13 | 1496 | 85 | |
AK027516.1 | - | 12 - 13 | 1345 | 85 | |
AK315484.1 | - | 12 - 13 | 1408 | 85 | |
BC001024.2 | - | 12 - 13 | 1343 | 85 |