Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFHX4
Entrez Gene ID
79776
Gene Name
zinc finger homeobox 4
Gene Aliases
ZFH4, ZHF4
Chromosome Location
Chr.8: 76681219 - 76867285 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024721.4 NP_078997.4 9 - 10 4347 71
XM_011517592.2 XP_011515894.1 9 - 10 4107 71
XM_011517593.2 XP_011515895.1 10 - 11 4101 71
XM_011517594.2 XP_011515896.1 9 - 10 4112 71
XM_011517595.2 XP_011515897.1 9 - 10 4181 71
XM_011517596.2 XP_011515898.1 9 - 10 4330 71
XM_011517597.2 XP_011515899.1 8 - 9 4291 71
XM_017013845.1 XP_016869334.1 8 - 9 4213 71
GenBank mRNA AB083343.2 - 10 - 11 4339 71
AK095274.1 - 9 - 10 2966 71
AY260762.1 - 8 - 9 3869 71

View Full Product Details