Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAEL
Entrez Gene ID
84944
Gene Name
maelstrom spermatogenic transposon silencer
Gene Aliases
CT128, SPATA35
Chromosome Location
Chr.1: 166921121 - 167022214 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286377.1 NP_001273306.1 3 - 4 656 115
NM_001286378.1 NP_001273307.1 5 - 6 522 115
NM_032858.2 NP_116247.1 4 - 5 749 115
XM_006711583.1 XP_006711646.1 5 - 6 553 115
XM_011510067.1 XP_011508369.1 4 - 5 715 115
XM_011510068.1 XP_011508370.1 5 - 6 506 115
XM_017002602.1 XP_016858091.1 6 - 7 890 115
XM_017002603.1 XP_016858092.1 5 - 6 552 115
GenBank mRNA AK027810.1 - 4 - 5 556 115
AK131066.1 - 4 - 5 559 115
AK302253.1 - 3 - 4 472 115
AL133073.1 - 3 - 4 313 115
BC028595.2 - 4 - 5 529 115
BC034310.1 - 3 - 4 1034 115
DQ076156.2 - 4 - 5 558 115
GU383113.1 - 3 - 4 465 115
GU383114.1 - 5 - 6 522 115

View Full Product Details