The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286377.1 | NP_001273306.1 | 4 - 5 | 695 | 87 |
NM_001286378.1 | NP_001273307.1 | 6 - 7 | 561 | 87 | |
NM_032858.2 | NP_116247.1 | 5 - 6 | 788 | 87 | |
XM_006711583.1 | XP_006711646.1 | 6 - 7 | 592 | 87 | |
XM_011510067.1 | XP_011508369.1 | 5 - 6 | 754 | 87 | |
XM_011510068.1 | XP_011508370.1 | 6 - 7 | 545 | 87 | |
XM_017002602.1 | XP_016858091.1 | 7 - 8 | 929 | 87 | |
XM_017002603.1 | XP_016858092.1 | 6 - 7 | 591 | 87 | |
GenBank mRNA | AK027810.1 | - | 5 - 6 | 595 | 87 |
AK131066.1 | - | 5 - 6 | 598 | 87 | |
AK302253.1 | - | 4 - 5 | 511 | 87 | |
AL133073.1 | - | 4 - 5 | 352 | 87 | |
BC028595.2 | - | 5 - 6 | 568 | 87 | |
BC034310.1 | - | 4 - 5 | 1073 | 87 | |
DQ076156.2 | - | 5 - 6 | 597 | 87 | |
GU383113.1 | - | 4 - 5 | 504 | 87 | |
GU383114.1 | - | 6 - 7 | 561 | 87 |