Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAEL
Entrez Gene ID
84944
Gene Name
maelstrom spermatogenic transposon silencer
Gene Aliases
CT128, SPATA35
Chromosome Location
Chr.1: 166921121 - 167022214 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286377.1 NP_001273306.1 5 - 6 822 72
NM_001286378.1 NP_001273307.1 7 - 8 688 72
NM_032858.2 NP_116247.1 6 - 7 915 72
XM_006711583.1 XP_006711646.1 7 - 8 719 72
XM_011510067.1 XP_011508369.1 6 - 7 881 72
XM_011510068.1 XP_011508370.1 7 - 8 672 72
XM_017002602.1 XP_016858091.1 8 - 9 1056 72
XM_017002603.1 XP_016858092.1 7 - 8 718 72
GenBank mRNA AK027810.1 - 6 - 7 722 72
AK131066.1 - 6 - 7 725 72
AK302253.1 - 5 - 6 638 72
AL133073.1 - 5 - 6 479 72
BC028595.2 - 6 - 7 695 72
BC034310.1 - 5 - 6 1200 72
DQ076156.2 - 6 - 7 724 72
GU383113.1 - 5 - 6 631 72
GU383114.1 - 7 - 8 688 72

View Full Product Details