The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013031.2 | NP_001013049.1 | 14 - 15 | 2119 | 59 |
NM_001206569.1 | NP_001193498.1 | 14 - 15 | 2119 | 59 | |
NM_001206570.1 | NP_001193499.1 | 14 - 15 | 2119 | 59 | |
NM_001206571.1 | NP_001193500.1 | 14 - 15 | 2119 | 59 | |
NM_001206572.1 | NP_001193501.1 | 14 - 15 | 2119 | 59 | |
NM_052918.4 | NP_443150.3 | 14 - 15 | 2119 | 59 | |
XM_011539199.2 | XP_011537501.1 | 14 - 15 | 1976 | 59 | |
XM_011539201.2 | XP_011537503.1 | 14 - 15 | 1976 | 59 | |
XM_017015614.1 | XP_016871103.1 | 14 - 15 | 1976 | 59 | |
XM_017015615.1 | XP_016871104.1 | 14 - 15 | 1976 | 59 | |
XM_017015616.1 | XP_016871105.1 | 14 - 15 | 1655 | 59 | |
XM_017015617.1 | XP_016871106.1 | 14 - 15 | 1454 | 59 | |
XM_017015618.1 | XP_016871107.1 | 6 - 7 | 610 | 59 | |
GenBank mRNA | AB209142.1 | - | 14 - 15 | 1979 | 59 |
AK123752.1 | - | 6 - 7 | 673 | 59 | |
AK125464.1 | - | 14 - 15 | 2119 | 59 | |
AK126905.1 | - | 14 - 15 | 1483 | 59 | |
AK289797.1 | - | 14 - 15 | 2120 | 59 | |
BC131597.1 | - | 14 - 15 | 1945 | 59 |