Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORCS1
Entrez Gene ID
114815
Gene Name
sortilin related VPS10 domain containing receptor 1
Gene Aliases
hSorCS
Chromosome Location
Chr.10: 106573663 - 107181138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013031.2 NP_001013049.1 23 - 24 3349 67
NM_001206569.1 NP_001193498.1 23 - 24 3349 67
NM_001206570.1 NP_001193499.1 23 - 24 3349 67
NM_001206571.1 NP_001193500.1 23 - 24 3349 67
NM_001206572.1 NP_001193501.1 23 - 24 3349 67
NM_052918.4 NP_443150.3 23 - 24 3349 67
XM_011539199.2 XP_011537501.1 23 - 24 3206 67
XM_011539201.2 XP_011537503.1 23 - 24 3206 67
XM_017015614.1 XP_016871103.1 23 - 24 3206 67
XM_017015616.1 XP_016871105.1 23 - 24 2885 67
XM_017015617.1 XP_016871106.1 23 - 24 2684 67
XM_017015618.1 XP_016871107.1 15 - 16 1840 67
GenBank mRNA AB209142.1 - 23 - 24 3209 67
AF284756.1 - 23 - 24 3175 67
AK074581.1 - 1 - 2 55 67
AK123752.1 - 15 - 16 1903 67
AK125464.1 - 23 - 24 3349 67
AK126905.1 - 23 - 24 2713 67
AK289797.1 - 23 - 24 3350 67
AY099452.1 - 23 - 24 3175 67
AY099453.1 - 23 - 24 3175 67
BC131597.1 - 23 - 24 3175 67
EU281343.1 - 23 - 24 3175 67
EU281344.1 - 23 - 24 3175 67

View Full Product Details