Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORCS1
Entrez Gene ID
114815
Gene Name
sortilin related VPS10 domain containing receptor 1
Gene Aliases
hSorCS
Chromosome Location
Chr.10: 106573663 - 107181138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013031.2 NP_001013049.1 8 - 9 1417 68
NM_001206569.1 NP_001193498.1 8 - 9 1417 68
NM_001206570.1 NP_001193499.1 8 - 9 1417 68
NM_001206571.1 NP_001193500.1 8 - 9 1417 68
NM_001206572.1 NP_001193501.1 8 - 9 1417 68
NM_052918.4 NP_443150.3 8 - 9 1417 68
XM_011539199.2 XP_011537501.1 8 - 9 1274 68
XM_011539201.2 XP_011537503.1 8 - 9 1274 68
XM_017015614.1 XP_016871103.1 8 - 9 1274 68
XM_017015615.1 XP_016871104.1 8 - 9 1274 68
XM_017015616.1 XP_016871105.1 8 - 9 953 68
XM_017015617.1 XP_016871106.1 8 - 9 752 68
GenBank mRNA AB209142.1 - 8 - 9 1277 68
AF284756.1 - 8 - 9 1243 68
AK125464.1 - 8 - 9 1417 68
AK126905.1 - 8 - 9 781 68
AK289797.1 - 8 - 9 1418 68
AY099452.1 - 8 - 9 1243 68
AY099453.1 - 8 - 9 1243 68
BC131597.1 - 8 - 9 1243 68
EU281343.1 - 8 - 9 1243 68
EU281344.1 - 8 - 9 1243 68

View Full Product Details