Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMC3
Entrez Gene ID
9126
Gene Name
structural maintenance of chromosomes 3
Gene Aliases
BAM, BMH, CDLS3, CSPG6, HCAP, SMC3L1
Chromosome Location
Chr.10: 110567691 - 110604634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005445.3 NP_005436.1 20 - 21 2391 66
GenBank mRNA AF020043.1 - 20 - 21 2356 66
AF067163.1 - 8 - 9 1039 66
AK289771.1 - 20 - 21 2387 66
AK315845.1 - 20 - 21 2390 66
BC047324.1 - 20 - 21 2286 66

View Full Product Details