The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005445.3 | NP_005436.1 | 28 - 29 | 3697 | 82 |
GenBank mRNA | AF020043.1 | - | 28 - 29 | 3662 | 82 |
AF067163.1 | - | 16 - 17 | 2345 | 82 | |
AJ005015.1 | - | 7 - 8 | 1095 | 82 | |
AK289771.1 | - | 28 - 29 | 3693 | 82 | |
AK315845.1 | - | 28 - 29 | 3696 | 82 | |
BC047324.1 | - | 28 - 29 | 3592 | 82 |