The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012967.2 | NP_001012985.2 | 2 - 3 | 253 | 100 |
NM_001291510.1 | NP_001278439.1 | 2 - 3 | 253 | 100 | |
XM_005263341.4 | XP_005263398.2 | 2 - 3 | 236 | 100 | |
XM_005263342.2 | XP_005263399.2 | 2 - 3 | 253 | 100 | |
XM_005263343.2 | XP_005263400.2 | 2 - 3 | 219 | 100 | |
XM_005263345.4 | XP_005263402.2 | 2 - 3 | 232 | 100 | |
XM_006714408.2 | XP_006714471.1 | 3 - 4 | 303 | 100 | |
XM_011532402.2 | XP_011530704.1 | 3 - 4 | 526 | 100 | |
XM_011532403.2 | XP_011530705.1 | 3 - 4 | 361 | 100 | |
XM_011532404.1 | XP_011530706.1 | 2 - 3 | 253 | 100 | |
XM_011532405.1 | XP_011530707.1 | 2 - 3 | 253 | 100 | |
XM_011532406.1 | XP_011530708.1 | 2 - 3 | 253 | 100 | |
XM_017008829.1 | XP_016864318.1 | 2 - 3 | 219 | 100 | |
XM_017008830.1 | XP_016864319.1 | 2 - 3 | 253 | 100 | |
GenBank mRNA | AK096369.1 | - | 3 - 4 | 303 | 100 |
CR933601.1 | - | 2 - 3 | 227 | 100 | |
DA553350.1 | - | 2 - 3 | 253 | 100 |