Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX60L
Entrez Gene ID
91351
Gene Name
DEAD-box helicase 60-like
Gene Aliases
-
Chromosome Location
Chr.4: 168356735 - 168480514 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012967.2 NP_001012985.2 33 - 34 4732 136
XM_005263341.4 XP_005263398.2 33 - 34 4718 136
XM_005263342.2 XP_005263399.2 33 - 34 4735 136
XM_005263343.2 XP_005263400.2 33 - 34 4701 136
XM_005263345.4 XP_005263402.2 33 - 34 4714 136
XM_006714408.2 XP_006714471.1 34 - 35 4785 136
XM_011532402.2 XP_011530704.1 34 - 35 5008 136
XM_011532403.2 XP_011530705.1 34 - 35 4843 136
XM_011532404.1 XP_011530706.1 32 - 33 4639 136
XM_011532405.1 XP_011530707.1 32 - 33 4636 136
XM_011532406.1 XP_011530708.1 32 - 33 4630 136
XM_011532409.2 XP_011530711.1 21 - 22 2775 136
XM_017008829.1 XP_016864318.1 32 - 33 4605 136
XM_017008830.1 XP_016864319.1 32 - 33 4582 136
XM_017008831.1 XP_016864320.1 22 - 23 3037 136
GenBank mRNA AK023743.1 - 12 - 13 1619 136
AL832950.1 - 14 - 15 1827 136
KR632542.1 - 31 - 32 4484 136

View Full Product Details