The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012967.2 | NP_001012985.2 | 35 - 36 | 5024 | 122 |
XM_005263341.4 | XP_005263398.2 | 35 - 36 | 5010 | 122 | |
XM_005263342.2 | XP_005263399.2 | 35 - 36 | 5027 | 122 | |
XM_005263343.2 | XP_005263400.2 | 35 - 36 | 4993 | 122 | |
XM_005263345.4 | XP_005263402.2 | 35 - 36 | 5006 | 122 | |
XM_006714408.2 | XP_006714471.1 | 36 - 37 | 5077 | 122 | |
XM_011532402.2 | XP_011530704.1 | 36 - 37 | 5300 | 122 | |
XM_011532403.2 | XP_011530705.1 | 36 - 37 | 5135 | 122 | |
XM_011532404.1 | XP_011530706.1 | 34 - 35 | 4931 | 122 | |
XM_011532405.1 | XP_011530707.1 | 34 - 35 | 4928 | 122 | |
XM_011532406.1 | XP_011530708.1 | 34 - 35 | 4922 | 122 | |
XM_011532409.2 | XP_011530711.1 | 23 - 24 | 3067 | 122 | |
XM_017008829.1 | XP_016864318.1 | 34 - 35 | 4897 | 122 | |
XM_017008830.1 | XP_016864319.1 | 34 - 35 | 4874 | 122 | |
XM_017008831.1 | XP_016864320.1 | 24 - 25 | 3329 | 122 | |
GenBank mRNA | AK023530.1 | - | 6 - 7 | 1751 | 122 |
AK023743.1 | - | 14 - 15 | 1911 | 122 | |
AL832950.1 | - | 16 - 17 | 2119 | 122 | |
EB710202.1 | - | 3 - 4 | 375 | 122 | |
KR632542.1 | - | 33 - 34 | 4776 | 122 |