The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020870.3 | NP_065921.2 | 10 - 11 | 2481 | 66 |
GenBank mRNA | AB040927.1 | - | 6 - 7 | 1396 | 66 |
AK025255.1 | - | 6 - 7 | 1313 | 66 | |
BC053671.1 | - | 8 - 9 | 2823 | 66 |