The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020870.3 | NP_065921.2 | 4 - 5 | 1107 | 87 |
GenBank mRNA | AK021429.1 | - | 4 - 5 | 968 | 87 |
BC033203.1 | - | 3 - 4 | 654 | 87 | |
BC041023.1 | - | 2 - 3 | 357 | 87 | |
BC053671.1 | - | - | 945 | 87 |