The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040446.2 | NP_001035536.1 | 6 - 7 | 751 | 69 |
NM_001294343.1 | NP_001281272.1 | 6 - 7 | 751 | 69 | |
NM_001294344.1 | NP_001281273.1 | 6 - 7 | 751 | 69 | |
XM_005248313.3 | XP_005248370.1 | 6 - 7 | 715 | 69 | |
XM_011514057.1 | XP_011512359.1 | 6 - 7 | 715 | 69 | |
XM_011514058.2 | XP_011512360.1 | 6 - 7 | 715 | 69 | |
XM_011514059.1 | XP_011512361.1 | 6 - 7 | 715 | 69 | |
XM_011514060.1 | XP_011512362.1 | 6 - 7 | 715 | 69 | |
XM_017009589.1 | XP_016865078.1 | 6 - 7 | 715 | 69 | |
XM_017009590.1 | XP_016865079.1 | 7 - 8 | 653 | 69 | |
XM_017009591.1 | XP_016865080.1 | 5 - 6 | 664 | 69 | |
XM_017009592.1 | XP_016865081.1 | 4 - 5 | 455 | 69 | |
XM_017009593.1 | XP_016865082.1 | 5 - 6 | 642 | 69 | |
XM_017009594.1 | XP_016865083.1 | 6 - 7 | 703 | 69 | |
XM_017009595.1 | XP_016865084.1 | 4 - 5 | 585 | 69 | |
XM_017009596.1 | XP_016865085.1 | 6 - 7 | 639 | 69 | |
GenBank mRNA | AB051469.1 | - | 6 - 7 | 684 | 69 |
AY028703.1 | - | 6 - 7 | 688 | 69 | |
BC057393.1 | - | 6 - 7 | 681 | 69 |