The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170760.1 | NP_001164231.1 | 12 - 13 | 1436 | 92 |
NM_001170761.1 | NP_001164232.1 | 13 - 14 | 1337 | 92 | |
NM_014370.3 | NP_055185.2 | 12 - 13 | 1439 | 92 | |
GenBank mRNA | AF027406.1 | - | 13 - 14 | 1275 | 92 |
AK301749.1 | - | 12 - 13 | 1399 | 92 | |
AK314828.1 | - | 12 - 13 | 1399 | 92 | |
BC092416.1 | - | 12 - 13 | 1385 | 92 | |
BC117124.1 | - | 12 - 13 | 1430 | 92 | |
DQ099381.1 | - | 12 - 13 | 1350 | 92 | |
U82808.1 | - | 13 - 14 | 1155 | 92 |