The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170760.1 | NP_001164231.1 | 7 - 8 | 833 | 146 |
NM_001170761.1 | NP_001164232.1 | 7 - 8 | 833 | 146 | |
GenBank mRNA | AF027406.1 | - | 7 - 8 | 771 | 146 |
BC117124.1 | - | 7 - 8 | 827 | 146 | |
DQ099381.1 | - | 7 - 8 | 747 | 146 | |
U82808.1 | - | 7 - 8 | 651 | 146 |