The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018412.3 | NP_060882.2 | 2 - 3 | 441 | 111 |
NM_021908.2 | NP_068708.1 | 2 - 3 | 441 | 111 | |
GenBank mRNA | AF234882.1 | - | 2 - 3 | 440 | 111 |
AF234883.1 | - | 2 - 3 | 440 | 111 | |
AJ277291.1 | - | 1 - 2 | 76 | 111 | |
AK093212.1 | - | 3 - 4 | 690 | 111 | |
AK292372.1 | - | 2 - 3 | 267 | 111 | |
AK297510.1 | - | 2 - 3 | 318 | 111 | |
AK297861.1 | - | 2 - 3 | 149 | 111 | |
AK298540.1 | - | 2 - 3 | 263 | 111 | |
AY009152.1 | - | 2 - 3 | 432 | 111 | |
AY009153.1 | - | 2 - 3 | 432 | 111 | |
BC030954.1 | - | 2 - 3 | 267 | 111 | |
BC075855.1 | - | 2 - 3 | 441 | 111 | |
KJ534947.1 | - | 2 - 3 | 227 | 111 |