The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321298.1 | NP_001308227.1 | 6 - 7 | 978 | 86 |
NM_173637.3 | NP_775908.2 | 5 - 6 | 880 | 86 | |
NR_135612.1 | 5 - 6 | 880 | 86 | ||
XM_011527926.1 | XP_011526228.1 | 5 - 6 | 674 | 86 | |
GenBank mRNA | AK097761.1 | - | 6 - 7 | 962 | 86 |
BC031671.1 | - | 5 - 6 | 880 | 86 |