Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CDK16
- Entrez Gene ID
-
5127
- Gene Name
- cyclin dependent kinase 16
- Gene Aliases
-
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
- Chromosome Location
-
Chr.X: 47217881 - 47229997 on Build GRCh38
View Full Product Details