The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204466.1 | NP_001191395.1 | 17 - 18 | 2259 | 68 |
NM_001204467.1 | NP_001191396.1 | 18 - 19 | 2487 | 68 | |
NM_001204468.1 | NP_001191397.1 | 18 - 19 | 2521 | 68 | |
NM_005676.4 | NP_005667.2 | 18 - 19 | 2490 | 68 | |
NM_152856.2 | NP_690595.1 | 17 - 18 | 2256 | 68 | |
XM_005272677.4 | XP_005272734.1 | 18 - 19 | 2540 | 68 | |
XM_005272678.4 | XP_005272735.1 | 17 - 18 | 2074 | 68 | |
XM_005272679.4 | XP_005272736.1 | 17 - 18 | 2071 | 68 | |
XM_006724563.1 | XP_006724626.1 | 14 - 15 | 1707 | 68 | |
XM_011543989.2 | XP_011542291.1 | 10 - 11 | 1431 | 68 | |
XM_017029884.1 | XP_016885373.1 | 17 - 18 | 2410 | 68 | |
XM_017029885.1 | XP_016885374.1 | 17 - 18 | 2182 | 68 | |
GenBank mRNA | AK024839.1 | - | 1 - 2 | 98 | 68 |
AK025854.1 | - | 6 - 7 | 863 | 68 | |
AK098115.1 | - | - | 1825 | 68 | |
AK292758.1 | - | 17 - 18 | 2210 | 68 | |
AL137421.1 | - | 19 - 20 | 2379 | 68 | |
BC003089.1 | - | 17 - 18 | 2195 | 68 | |
BC004181.2 | - | 18 - 19 | 2443 | 68 | |
BC008733.2 | - | 18 - 19 | 2475 | 68 | |
BC024153.2 | - | 17 - 18 | 2105 | 68 | |
BX537969.1 | - | 18 - 19 | 2472 | 68 | |
D50912.1 | - | 18 - 19 | 2338 | 68 | |
KU178411.1 | - | 16 - 17 | 1863 | 68 | |
KU178412.1 | - | 17 - 18 | 2097 | 68 | |
U35373.1 | - | 4 - 5 | 72 |