The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204466.1 | NP_001191395.1 | 3 - 4 | 591 | 89 |
NM_152856.2 | NP_690595.1 | 3 - 4 | 591 | 89 | |
XM_005272678.4 | XP_005272735.1 | 3 - 4 | 406 | 89 | |
XM_005272679.4 | XP_005272736.1 | 3 - 4 | 406 | 89 | |
XM_006724563.1 | XP_006724626.1 | 1 - 2 | 113 | 89 | |
GenBank mRNA | AK098115.1 | - | - | 113 | 89 |
AK292758.1 | - | 3 - 4 | 542 | 89 | |
BC003089.1 | - | 3 - 4 | 530 | 89 | |
KU178411.1 | - | 2 - 3 | 198 | 89 |