Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAMLD1
Entrez Gene ID
10046
Gene Name
mastermind like domain containing 1
Gene Aliases
CG1, CXorf6, F18, HYSP2
Chromosome Location
Chr.X: 150361422 - 150514178 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177466.2 NP_001170937.1 3 - 4 2034 141
NM_005491.4 NP_005482.2 4 - 5 2109 141
XM_006724802.3 XP_006724865.1 6 - 7 2457 141
XM_006724803.3 XP_006724866.1 5 - 6 2382 141
XM_017029188.1 XP_016884677.1 6 - 7 2329 141
GenBank mRNA AK302612.1 - 6 - 7 1881 141
AK315217.1 - 4 - 5 1982 141
BC136324.1 - 3 - 4 2019 141
DA527328.1 - 2 - 3 188 141
U46023.1 - 4 - 5 2110 141

View Full Product Details