The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013328.3 | NP_037460.2 | 3 - 4 | 550 | 74 |
GenBank mRNA | AF087859.1 | - | 3 - 4 | 468 | 74 |
AK291913.1 | - | 3 - 4 | 385 | 74 | |
AL834217.1 | - | 1 - 2 | 547 | 74 | |
BC014868.1 | - | 3 - 4 | 441 | 74 | |
BC020553.1 | - | 3 - 4 | 475 | 74 | |
KU178615.1 | - | 3 - 4 | 320 | 74 |