The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323930.1 | NP_001310859.1 | 2 - 3 | 805 | 70 |
NM_001323931.1 | NP_001310860.1 | 3 - 4 | 1137 | 70 | |
NM_001323932.1 | NP_001310861.1 | 3 - 4 | 968 | 70 | |
NM_001323933.1 | NP_001310862.1 | 4 - 5 | 913 | 70 | |
NM_001323934.1 | NP_001310863.1 | 2 - 3 | 572 | 70 | |
NM_001323935.1 | NP_001310864.1 | 2 - 3 | 1040 | 70 | |
NM_053052.3 | NP_444280.2 | 2 - 3 | 1040 | 70 | |
NR_136655.1 | 2 - 3 | 1040 | 70 | ||
XM_006711734.3 | XP_006711797.1 | 2 - 3 | 846 | 70 | |
XM_017000231.1 | XP_016855720.1 | 4 - 5 | 4079 | 70 | |
XM_017000232.1 | XP_016855721.1 | 3 - 4 | 3885 | 70 | |
GenBank mRNA | AK054633.1 | - | 2 - 3 | 600 | 70 |
AY090635.2 | - | 2 - 3 | 1040 | 70 | |
BC001332.1 | - | 1 - 2 | 379 | 70 | |
BC004418.1 | - | 1 - 2 | 370 | 70 | |
BC007786.2 | - | 2 - 3 | 583 | 70 | |
BC011145.1 | - | 2 - 3 | 593 | 70 | |
BC014059.2 | - | 1 - 2 | 541 | 70 | |
BC018760.1 | - | 2 - 3 | 879 | 70 | |
FJ236305.1 | - | 1 - 2 | 541 | 70 | |
GQ472206.1 | - | 2 - 3 | 583 | 70 |