The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177515.1 | NP_001170986.1 | 10 - 11 | 1289 | 72 |
NM_021160.2 | NP_066983.1 | 12 - 13 | 1162 | 72 | |
NR_033488.1 | 12 - 13 | 1413 | 72 | ||
NR_033489.1 | 10 - 11 | 1038 | 72 | ||
GenBank mRNA | AF193047.2 | - | 12 - 13 | 1111 | 72 |
AK001207.1 | - | 7 - 8 | 609 | 72 | |
AK023194.1 | - | 12 - 13 | 1113 | 72 | |
AK055255.1 | - | 12 - 13 | 1413 | 72 | |
AK289469.1 | - | 12 - 13 | 1158 | 72 | |
AK297712.1 | - | 10 - 11 | 1286 | 72 | |
BC031839.1 | - | 12 - 13 | 1117 | 72 |