The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144506.2 | NP_653089.1 | 2 - 3 | 244 | 66 |
GenBank mRNA | BM845575.1 | - | 2 - 3 | 210 | 66 |