Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIN2C
Entrez Gene ID
2905
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
Gene Aliases
GluN2C, NMDAR2C, NR2C
Chromosome Location
Chr.17: 74842021 - 74860843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000835.4 NP_000826.2 12 - 13 2734 67
NR_103735.1 12 - 13 2730 67
XM_006721845.3 XP_006721908.1 11 - 12 4000 67
XM_006721846.3 XP_006721909.1 11 - 12 3998 67
XM_011524686.2 XP_011522988.1 12 - 13 4089 67
XM_011524687.2 XP_011522989.1 12 - 13 4065 67
XM_011524688.2 XP_011522990.1 11 - 12 4025 67
XM_011524689.2 XP_011522991.1 13 - 14 2812 67
XM_017024544.1 XP_016880033.1 12 - 13 4090 67
GenBank mRNA AB208799.1 - 8 - 9 6323 67
BC140801.1 - 12 - 13 2612 67
L76224.1 - 11 - 12 2578 67
U77782.1 - 12 - 13 2766 67

View Full Product Details