Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCGF1
Entrez Gene ID
84759
Gene Name
polycomb group ring finger 1
Gene Aliases
2010002K04Rik, NSPC1, RNF3A-2, RNF68
Chromosome Location
Chr.2: 74505043 - 74508580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032673.2 NP_116062.2 3 - 4 376 75
GenBank mRNA AF087884.1 - 2 - 3 482 75
AK125742.1 - 3 - 4 1262 75
BC004952.1 - 3 - 4 341 75
BU602498.1 - 3 - 4 379 75

View Full Product Details