The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024605.3 | NP_078881.3 | 14 - 15 | 1301 | 69 |
XM_005263215.3 | XP_005263272.1 | 14 - 15 | 1537 | 69 | |
XM_017008602.1 | XP_016864091.1 | 15 - 16 | 1573 | 69 | |
GenBank mRNA | AB050785.1 | - | 14 - 15 | 1301 | 69 |
AK123034.1 | - | 5 - 6 | 333 | 69 | |
AK304458.1 | - | 6 - 7 | 484 | 69 | |
BC011920.2 | - | 10 - 11 | 831 | 69 | |
BC109029.2 | - | 14 - 15 | 1291 | 69 | |
BC109030.1 | - | 14 - 15 | 1290 | 69 | |
BC126899.1 | - | 14 - 15 | 1297 | 69 | |
BC128055.1 | - | 14 - 15 | 1301 | 69 | |
BX647727.1 | - | 8 - 9 | 6431 | 69 |