Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYN3
Entrez Gene ID
8224
Gene Name
synapsin III
Gene Aliases
-
Chromosome Location
Chr.22: 32508022 - 33058391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135774.1 NP_001129246.1 2 - 3 571 89
NM_003490.3 NP_003481.3 1 - 2 468 89
NM_133633.2 NP_598344.2 1 - 2 468 89
XM_011530405.2 XP_011528707.1 2 - 3 1104 89
XM_011530406.2 XP_011528708.1 2 - 3 570 89
XM_011530407.2 XP_011528709.1 2 - 3 977 89
XM_011530408.1 XP_011528710.1 2 - 3 568 89
XM_017028960.1 XP_016884449.1 3 - 4 1502 89
XM_017028961.1 XP_016884450.1 2 - 3 894 89
XM_017028962.1 XP_016884451.1 2 - 3 829 89
XM_017028963.1 XP_016884452.1 2 - 3 493 89
XM_017028964.1 XP_016884453.1 2 - 3 568 89
GenBank mRNA AB209684.1 - 2 - 3 523 89
AF046873.1 - 1 - 2 345 89
BC075065.2 - 1 - 2 335 89
BC075066.2 - 1 - 2 335 89
BC117460.1 - 1 - 2 336 89
BC143874.1 - 1 - 2 336 89
CR456589.1 - 1 - 2 400 89
DQ655947.2 - 2 - 3 795 89

View Full Product Details