The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004914.3 | NP_004905.2 | 4 - 5 | 464 | 83 |
XM_005261859.4 | XP_005261916.1 | 5 - 6 | 548 | 83 | |
XM_006724381.3 | XP_006724444.1 | 5 - 6 | 551 | 83 | |
XM_006724382.3 | XP_006724445.1 | 4 - 5 | 476 | 83 | |
XM_011530544.2 | XP_011528846.1 | 5 - 6 | 556 | 83 | |
XM_011530545.2 | XP_011528847.1 | 5 - 6 | 552 | 83 | |
XM_011530546.2 | XP_011528848.1 | 5 - 6 | 558 | 83 | |
XM_011530547.2 | XP_011528849.1 | 3 - 4 | 398 | 83 | |
XM_011530550.2 | XP_011528852.1 | 5 - 6 | 562 | 83 | |
XM_017029104.1 | XP_016884593.1 | 4 - 5 | 472 | 83 | |
XM_017029105.1 | XP_016884594.1 | 3 - 4 | 394 | 83 | |
XM_017029106.1 | XP_016884595.1 | 3 - 4 | 276 | 83 | |
XM_017029107.1 | XP_016884596.1 | 4 - 5 | 312 | 83 | |
GenBank mRNA | AB023061.1 | - | 4 - 5 | 464 | 83 |
AF133588.1 | - | 4 - 5 | 434 | 83 | |
BC104989.1 | - | 4 - 5 | 453 | 83 | |
BC104991.1 | - | 4 - 5 | 453 | 83 | |
CR456553.1 | - | 4 - 5 | 442 | 83 |