The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122819.2 | NP_001116291.1 | 13 - 14 | 78 | |
NM_001287212.1 | NP_001274141.1 | 13 - 14 | 78 | ||
NM_020816.3 | NP_065867.2 | 13 - 14 | 78 | ||
XM_005245950.3 | XP_005246007.1 | 13 - 14 | 78 | ||
XM_005245951.3 | XP_005246008.1 | 13 - 14 | 78 | ||
XM_011541837.1 | XP_011540139.1 | 13 - 14 | 78 | ||
XM_011541838.1 | XP_011540140.1 | 13 - 14 | 78 | ||
XM_011541839.2 | XP_011540141.1 | 13 - 14 | 78 | ||
XM_011541840.2 | XP_011540142.1 | 13 - 14 | 78 | ||
XM_011541841.2 | XP_011540143.1 | 13 - 14 | 78 | ||
XM_011541842.1 | XP_011540144.1 | 13 - 14 | 78 | ||
XM_011541843.1 | XP_011540145.1 | 13 - 14 | 78 | ||
XM_011541844.2 | XP_011540146.1 | 13 - 14 | 78 | ||
XM_011541845.2 | XP_011540147.1 | 13 - 14 | 78 | ||
XM_011541846.2 | XP_011540148.1 | 13 - 14 | 78 | ||
XM_011541847.2 | XP_011540149.1 | 7 - 8 | 78 | ||
GenBank mRNA | AB037826.2 | - | 13 - 14 | 2702 | 78 |
AB290162.1 | - | 13 - 14 | 2908 | 78 | |
AF009624.1 | - | 3 - 4 | 431 | 78 | |
AY484427.1 | - | 13 - 14 | 3100 | 78 | |
BC036871.1 | - | 4 - 5 | 940 | 78 | |
BC065927.1 | - | 13 - 14 | 2861 | 78 |