The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003800.4 | NP_003791.3 | 11 - 12 | 1494 | 95 |
XM_017011401.1 | XP_016866890.1 | 11 - 12 | 1581 | 95 | |
XM_017011402.1 | XP_016866891.1 | 11 - 12 | 1581 | 95 | |
GenBank mRNA | AB009022.1 | - | 11 - 12 | 1274 | 95 |
AB012142.1 | - | 11 - 12 | 1429 | 95 | |
AB012143.1 | - | 11 - 12 | 1429 | 95 | |
AF025654.1 | - | 11 - 12 | 1562 | 95 | |
AK295719.1 | - | 12 - 13 | 1372 | 95 | |
AK312407.1 | - | 11 - 12 | 1465 | 95 | |
BC019954.1 | - | 11 - 12 | 1461 | 95 |