Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RBM23
- Entrez Gene ID
-
55147
- Gene Name
- RNA binding motif protein 23
- Gene Aliases
-
CAPERbeta, PP239, RNPC4
- Chromosome Location
-
Chr.14: 22900645 - 22919187 on Build GRCh38
View Full Product Details