The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002830.3 | NP_002821.1 | 11 - 12 | 1600 | 155 |
XM_011511560.2 | XP_011509862.1 | 11 - 12 | 1505 | 155 | |
XM_011511562.2 | XP_011509864.1 | 10 - 11 | 1439 | 155 | |
XM_017004596.1 | XP_016860085.1 | 11 - 12 | 1526 | 155 | |
XM_017004597.1 | XP_016860086.1 | 11 - 12 | 1032 | 155 | |
XM_017004598.1 | XP_016860087.1 | 11 - 12 | 1505 | 155 | |
XM_017004599.1 | XP_016860088.1 | 11 - 12 | 1011 | 155 | |
XM_017004600.1 | XP_016860089.1 | 11 - 12 | 990 | 155 | |
XM_017004601.1 | XP_016860090.1 | 10 - 11 | 1481 | 155 | |
XM_017004602.1 | XP_016860091.1 | 10 - 11 | 1460 | 155 | |
XM_017004603.1 | XP_016860092.1 | 10 - 11 | 945 | 155 | |
XM_017004604.1 | XP_016860093.1 | 11 - 12 | 1484 | 155 | |
XM_017004606.1 | XP_016860095.1 | 11 - 12 | 1505 | 155 | |
GenBank mRNA | AK314836.1 | - | 11 - 12 | 1412 | 155 |
BC010674.1 | - | 11 - 12 | 1256 | 155 | |
M68941.1 | - | 11 - 12 | 1600 | 155 |