Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EYA2
Entrez Gene ID
2139
Gene Name
EYA transcriptional coactivator and phosphatase 2
Gene Aliases
EAB1
Chromosome Location
Chr.20: 46894624 - 47188844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005244.4 NP_005235.3 10 - 11 1350 133
NM_172110.3 NP_742108.2 10 - 11 1350 133
XM_005260327.2 XP_005260384.1 8 - 9 797 133
GenBank mRNA AF055015.1 - 6 - 7 664 133
AK293032.1 - 10 - 11 1054 133
BC000289.2 - 4 - 5 502 133
BC008803.2 - 10 - 11 1032 133
BC013882.2 - 10 - 11 1350 133
BT006682.1 - 9 - 10 904 133
U71207.1 - 9 - 10 978 133
U81601.1 - 1 - 2 94 133

View Full Product Details