Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBAC1
Entrez Gene ID
10422
Gene Name
UBA domain containing 1
Gene Aliases
GBDR1, UBADC1
Chromosome Location
Chr.9: 135932969 - 135961380 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016172.2 NP_057256.2 8 - 9 1181 120
GenBank mRNA AF038200.1 - 7 - 8 746 120
AF068195.1 - 7 - 8 808 120
AF176796.1 - 8 - 9 1151 120
BC004967.1 - 8 - 9 1181 120
BC011822.1 - 8 - 9 1007 120
BX647463.1 - 1 - 2 5234 120

View Full Product Details