The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114636.1 | NP_001108108.1 | 10 - 11 | 906 | 103 |
NM_018062.3 | NP_060532.2 | 10 - 11 | 891 | 103 | |
XM_005264395.3 | XP_005264452.1 | 10 - 11 | 891 | 103 | |
XM_005264397.4 | XP_005264454.1 | 6 - 7 | 610 | 103 | |
XM_011532939.2 | XP_011531241.1 | 11 - 12 | 951 | 103 | |
XM_011532940.2 | XP_011531242.1 | 11 - 12 | 936 | 103 | |
XM_011532941.1 | XP_011531243.1 | 11 - 12 | 951 | 103 | |
XM_011532942.2 | XP_011531244.1 | 10 - 11 | 906 | 103 | |
XM_011532944.2 | XP_011531246.1 | 7 - 8 | 628 | 103 | |
XM_011532945.2 | XP_011531247.1 | 6 - 7 | 596 | 103 | |
XM_017004414.1 | XP_016859903.1 | 11 - 12 | 936 | 103 | |
XM_017004416.1 | XP_016859905.1 | 6 - 7 | 610 | 103 | |
GenBank mRNA | AK001197.1 | - | 10 - 11 | 857 | 103 |
AK225147.1 | - | 9 - 10 | 747 | 103 | |
AK309671.1 | - | 7 - 8 | 706 | 103 | |
BC009042.1 | - | 10 - 11 | 861 | 103 | |
BC054517.1 | - | 10 - 11 | 866 | 103 |