The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024913.4 | NP_079189.4 | 18 - 19 | 2758 | 60 |
GenBank mRNA | AK289978.1 | - | 18 - 19 | 2755 | 60 |
AL832619.2 | - | 18 - 19 | 2713 | 60 | |
AL832638.2 | - | 18 - 19 | 2582 | 60 |