The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001406.3 | NP_001397.1 | 3 - 4 | 900 | 82 |
GenBank mRNA | AK314841.1 | - | 3 - 4 | 1006 | 82 |
BC022499.1 | - | 2 - 3 | 339 | 82 | |
BC042944.2 | - | 3 - 4 | 900 | 82 | |
U57001.1 | - | 3 - 4 | 900 | 82 | |
U66406.1 | - | 3 - 4 | 1116 | 82 |