The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303484.1 | NP_001290413.1 | 3 - 4 | 617 | 65 |
NM_031481.2 | NP_113669.1 | 4 - 5 | 680 | 65 | |
XM_011546149.2 | XP_011544451.1 | 4 - 5 | 1609 | 65 | |
XM_011546150.2 | XP_011544452.1 | 4 - 5 | 1479 | 65 | |
XM_011546151.2 | XP_011544453.1 | 4 - 5 | 868 | 65 | |
XM_011546152.1 | XP_011544454.1 | 4 - 5 | 495 | 65 | |
XM_011546153.2 | XP_011544455.1 | 4 - 5 | 1490 | 65 | |
XM_011546154.2 | XP_011544456.1 | 4 - 5 | 737 | 65 | |
XM_017028968.1 | XP_016884457.1 | 4 - 5 | 363 | 65 | |
XM_017028969.1 | XP_016884458.1 | 4 - 5 | 2509 | 65 | |
XM_017028970.1 | XP_016884459.1 | 5 - 6 | 2000 | 65 | |
XM_017028971.1 | XP_016884460.1 | 5 - 6 | 1989 | 65 | |
XM_017028972.1 | XP_016884461.1 | 5 - 6 | 1912 | 65 | |
XM_017028973.1 | XP_016884462.1 | 4 - 5 | 1937 | 65 | |
XM_017028974.1 | XP_016884463.1 | 5 - 6 | 1868 | 65 | |
XM_017028975.1 | XP_016884464.1 | 5 - 6 | 1780 | 65 | |
GenBank mRNA | AJ428203.1 | - | 4 - 5 | 636 | 65 |
AK095706.1 | - | 3 - 4 | 1602 | 65 | |
AK309110.1 | - | 3 - 4 | 464 | 65 | |
AY008285.1 | - | 4 - 5 | 636 | 65 | |
BC031644.1 | - | 3 - 4 | 572 | 65 | |
CR456578.1 | - | 2 - 3 | 266 | 65 |